missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Human IgG (H+L) Secondary Antibody, AP, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen PA130342
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-30342 detects Human IgG from human samples.
Human IgG (H+L) Polyclonal antibody specifically detects Human IgG (H+L) in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Spezifikation
Human IgG (H+L) | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Rabbit | |
IgG fraction | |
RUO | |
Human | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
0.05M tris HCl with 0.0001M ZnCl2, 0.001M MgCl2, 0.15M NaCl, 10mg/mL BSA, 50% glycerol and 0.1% sodium azide; pH 8.0 | |
human IgG whole molecule | |
1 mg | |
Secondary | |
4° C, do not freeze | |
IgG |