missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Human IgG Fc gamma Secondary Antibody, Invitrogen™

Rabbit Polyclonal Secondary Antibody

Marke:  Invitrogen 31142

Weitere Details : CAS : 17740-40-8 Netto-Gewicht : 0.00010kg

Artikelnummer. 10391944

  • 186.00 € / 2 Milligramm
Verfügbar ab: 10-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31142 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product #31142 reacts with the heavy chains of human IgG, but not with the light chains to most human immunoglobulins or with non-immunoglobulin serum proteins. This antibody does not react with human IgM or IgA. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. After opening, dilute only enough antibody for a single day's use. Store remainder at 4°C under sterile conditions. Country of Origin: USA

Human IgG Fc gamma Polyclonal antibody specifically detects Human IgG Fc gamma in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.
Spezifikation

Spezifikation

Human IgG Fc gamma
Polyclonal
Unconjugated
Rabbit
2 mg
Secondary
4° C
IgG
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
2.3 mg/mL
PBS with no preservative; pH 7.6
Antigen affinity chromatography
RUO
Human
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote