missing translation for 'onlineSavingsMsg'
Learn More

IgG (H+L) Rabbit anti-Human, FITC, Invitrogen™

Rabbit Polyclonal Secondary Antibody

Marke:  Invitrogen SA136099

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11545120

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beschreibung

Beschreibung

This antibody shows no cross reactivity with albumin fractions of human serum.

Thermo Scientific Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spezifikation

Spezifikation

Human IgG (H+L)
Polyclonal
FITC
Immunoglobulins.
1 mL
Secondary
4° C, store in dark
IgG
Immunohistochemistry
1.2 mg/mL
Rabbit
Purified
RUO
Human
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote