missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Human IgG (H+L) Cross-Adsorbed Secondary Antibody, Texas Red, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen PA128834
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Human IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Human IgG (H+L) Cross-Adsorbed in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.Spezifikation
Human IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Texas Red | |
Rabbit | |
IgG fraction | |
RUO | |
Human | |
Liquid |
Flow Cytometry, Immunocytochemistry | |
2 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl, 10mg/mL BSA and 0.01% sodium azide; pH 7.2 | |
human IgG whole molecule | |
100 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG |