Learn More
Rabbit anti-Mouse IgG F(ab')2 Secondary Antibody, AP, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen 31331
Weitere Details : Netto-Gewicht : 0.00100kg
Beschreibung
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. This antibody has been successfully used in Western blot and IP applications. Antibody Specificity: This antibody reacts with the light chains on mouse IgG and with those common to most mouse immunoglobulins, based on electrophoresis. The antibody does not react with the Fc portion of mouse IgG. No antibody was detected against non-immunoglobulin serum proteins. However, antibodies may cross-react with immunoglobulins from other species. Restoration and Storage: Store product at 4°C until opened. Restore with 1.0 mL distilled water (0.7 mg/mL after restoration). Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. To judge clarity, draw product into a pasteur pipette. Product is stable for several weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA
Mouse IgG F(ab')2 Polyclonal antibody specifically detects Mouse IgG F(ab')2 in Mouse samples. It is validated for Western Blot.Spezifikation
Mouse IgG F(ab')2 | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Rabbit | |
1 mL | |
Secondary | |
4° C | |
IgG |
Western Blot | |
0.7 mg/mL | |
TBS with 15mg/mL BSA and 0.05% sodium azide; pH 8 | |
Affinity chromatography | |
RUO | |
Mouse | |
Lyophilized |