Learn More
Rabbit anti-Mouse IgG F(ab')2 Secondary Antibody, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen 31192
Weitere Details : Netto-Gewicht : 0.80000kg
Beschreibung
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31192 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31192 reacts with the light chains of mouse IgG and with those common to other mouse immunoglobulins. This antibody does not react to the Fc portion of mouse IgG and non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. After opening, dilute only enough antibody for a single day’s use. Store remainder at 4°C under sterile conditions. Country of Origin: USA
Mouse IgG F(ab')2 Polyclonal antibody specifically detects Mouse IgG F(ab')2 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Spezifikation
Mouse IgG F(ab')2 | |
Polyclonal | |
Unconjugated | |
Rabbit | |
2 mg | |
Secondary | |
4° C | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
2.4 mg/mL | |
PBS with no preservative; pH 7.6 | |
Antigen affinity chromatography | |
RUO | |
Mouse | |
Liquid |