Learn More
Rabbit anti-Mouse IgG (H+L) Secondary Antibody, AP, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen 31329
Weitere Details : Netto-Gewicht : 1.81440kg
Beschreibung
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. This antibody has been successfully used in Western blot and IP applications. Antibody Specificity: This antibody reacts with the heavy chains of mouse IgG and with the light chains common to most mouse immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Restoration and Storage: Store product at 4°C until opened. Restore with 1.0 mL distilled water (0.6 mg/mL after restoration). Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. To judge clarity, draw product into a pasteur pipette. Product may be stored for several weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA
Mouse IgG (H+L) Polyclonal antibody specifically detects Mouse IgG (H+L) in Mouse samples. It is validated for Western Blot.Spezifikation
Mouse IgG (H+L) | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Rabbit | |
1 mL | |
Secondary | |
4° C | |
IgG |
Western Blot | |
0.6 mg/mL | |
TBS with 15mg/mL BSA and 0.05% sodium azide; pH 8 | |
Affinity chromatography | |
RUO | |
Mouse | |
Lyophilized |