Learn More
Rabbit anti-Mouse IgG, IgM (H+L) Secondary Antibody, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen 31198
Weitere Details : Netto-Gewicht : 0.80000kg
Beschreibung
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31198 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31198 reacts with the heavy chains of mouse IgG and IgM and with the light chains common to most mouse immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. After opening, dilute only enough antibody for a single day’s use. Store remainder at 4°C under sterile conditions. Country of Origin: USA
Mouse IgG, IgM (H+L) Polyclonal antibody specifically detects Mouse IgG, IgM (H+L) in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Spezifikation
Mouse IgG, IgM (H+L) | |
Polyclonal | |
Unconjugated | |
Rabbit | |
2 mg | |
Secondary | |
4° C | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
2.4 mg/mL | |
PBS with no preservative; pH 7.6 | |
Antigen affinity chromatography | |
RUO | |
Mouse | |
Liquid |