missing translation for 'onlineSavingsMsg'
Learn More

F(ab')2-Rabbit anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, FITC, Invitrogen™

F(ab')2-Rabbit Polyclonal Secondary Antibody

Marke:  Invitrogen PA184348

353.35 EUR gültig bis 2024-03-29
Benutzen Sie den Promotions-Code "21615" um Ihren Promotions-Preis zu erhalten.



Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11519150

  • 530.00 € / 1 Milligramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Some cross reactivity with mouse IgM and IgA is expected, as is cross reactivity with rat IgG. Cross reactivity with human serum proteins has been minimized by solid phase adsorption. F(ab')2 fragments were prepared by pepsin digestion of the IgG followed by a gel filtration step to remove the remaining intact IgG or Fc fragments. For Flow Cytometry, use 50 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. FITC conjugated Rabbit F(ab')2 anti Mouse IgG antibody recognizes all subclasses of mouse IgG.

Mouse IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgG (H+L) Cross-Adsorbed in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry.
Spezifikation

Spezifikation

Mouse IgG (H+L) Cross-Adsorbed
Polyclonal
FITC
Rabbit
Affinity chromatography
RUO
Mouse
Liquid
Flow Cytometry, Immunocytochemistry
1 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Purified murine IgG.
1 mg
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote