missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Mouse IgM Secondary Antibody, FITC, Invitrogen™

Rabbit Polyclonal Secondary Antibody

Marke:  Invitrogen 31557

151.34 EUR gültig bis 2024-03-29
Benutzen Sie den Promotions-Code "21615" um Ihren Promotions-Preis zu erhalten.



Weitere Details : Netto-Gewicht : 0.80000kg

Artikelnummer. 11859170

  • 227.00 € / 1500 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31557 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31557 reacts with the heavy chain portion of mouse IgM, but does not react against normal mouse IgG or non-immunoglobulin serum proteins. However, this antibody may cross-react with IgM from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Fluorescein Amax= 492 nm; Emax= 520 nm. Fluorophore/Protein: 8.2 μg/mg; ∽ 3 moles FITC per mole IgG (lot-dependent). Reconstitute with 1.1 mL of distilled water (1.4 mg/mL after restoration). Country of Origin: USA

Mouse IgM Polyclonal antibody specifically detects Mouse IgM in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation.
Spezifikation

Spezifikation

Mouse IgM
Polyclonal
FITC
Rabbit
1.5 mg
Secondary
4° C, store in dark
IgG
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
1.4 mg/mL
PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6
Affinity chromatography
RUO
Mouse
Lyophilized
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote