missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC12A3, Rabbit, AP, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant Human, Rat SLC12A3.
Marke: Abnova PAB28757.100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Spezifikation
SLC12A3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant Human, Rat SLC12A3. | |
PBS, 50% glycerol and 0.09% sodium azide | |
SLC12A3 | |
Recombinant protein corresponding to SLC12A3 N-terminus (amino acids 74-95). | |
100 μg | |
Primary | |
1 ug/mL of PAB28757 was sufficient for detection of SLC12A3 ,∼160kDa, in 10ug of rat kidney tissue lysate by colorimetric immunoblot analysis using Goat anti-rabbit IgG:HRP as the secondary antibody. | |
Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
Immunofluorescence, Western Blot | |
Alkaline Phosphatase (AP) | |
Immunofluorescence Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
FLJ96318/NCCT/TSC | |
Rabbit | |
Affinity purification | |
RUO | |
6559/54300 | |
Human, Mouse, Rat | |
Liquid |
For Research Use Only