missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ RAD9 Polyclonal Antibody
Beskrivelse
Immunogen sequence: AIFTIKDSLL DGHFVLATLS DTDSHSQDLG SPERHQPVPQ LQAHSTPHPD DFANDDIDSY MIAMETTIGN EGSRVLPSIS LSPGPQPPKS PGPHSEEEDE AEPSTVPGTP PPKKFRSLFF GSILAPV Highest antigen sequence indentity to the following orthologs: Rat - 74%, Mouse - 72%.
RAD9 is a component of the 9-1-1 cell cycle checkpoint response complex that plays a major role in DNA repair. The 9-1-1 complex is recruited to DNA lesions upon damage by the RAD17-replication factor C clamp loader complex. RAD9 acts as a sliding clamp platform on DNA for several proteins involved in long-patch base excision repair.
Tekniske data
Tekniske data
| Antigen | RAD9 |
| Anwendungen | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Klassifikation | Polyclonal |
| Konzentration | 0.7 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gen | RAD9A |
| Gen-Zugriffsnummer | Q99638 |
| Gen-Alias | Cell cycle checkpoint control protein RAD9A; chromatin-binding protein RAD9; DNA repair exonuclease rad9 homolog A; DNA repair protein RAD9; hRAD9; mRAD9; QtsA-19913; Rad9; RAD9 checkpoint clamp component A; RAD9 homolog A; RAD9 homolog A (S. pombe); Rad9a; Rad9-like protein; rCG47324-like; unnamed protein product; YD9934.02C; YDR217C |
| Gensymbole | RAD9A |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?