missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Raffinose-Hydrat, Extra Pure, SLR, Fisher Chemical
Click to view available options
Menge:
25 g
Verpackung:
Kunststoffpulverbehälter
Spezifikation
CAS | 512-69-6 |
Schmelzpunkt | 80°C |
Verpackung | Kunststoffpulverbehälter |
Summenformel | C18H32O16 |
MDL-Nummer | MFCD00006630 |
Menge | 25 g |
Synonym | raffinose, gossypose, d-+-raffinose, melitose, melitriose, d-raffinose, 6g-alpha-d-galactosylsucrose, unii-n5o3qu595m, raffinose 8ci, beta-d-fructofuranosyl alpha-d-galactopyranosyl-1->6-alpha-d-glucopyranoside |
InChI-Schlüssel | MUPFEKGTMRGPLJ-ZQSKZDJDSA-N |
SMILES | OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO[C@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O |
IUPAC-Name | (2S,3R,4S,5R,6R)-2-{[(2R,3S,4S,5R,6R)-6-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-3,4,5-trihydroxyoxan-2-yl]methoxy}-6-(hydroxymethyl)oxane-3,4,5-triol |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'