missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ramifenazon-d7, TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | Ramifenazone-d7 |
| CAS | 1330180-51-2 |
| Hinweise zur Reinheitsqualität | HPLC |
| Summenformel | C14 D7 H12 N3 O |
| Menge | 5 mg |
| Synonym | 1,2-Dihydro-1,5-dimethyl-4-[(1-methylethyl-d7)amino]-2-phenyl-3H-pyrazol-3-one, 4-(Isopropylamino-d7)antipyrine, 1-Phenyl-2,3-dimethyl-4-(isopropyl-d7) aminopyrazolone, 4-(Isopropylamino-d7)phenazone, 4-(Isopropyl-d7)amino- 2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, 4-Mono(isopropyl-d7)amino-1-phenyl- 2,3-dimethyl-5-pyrazolone, Isopirina-d7, (Isopropyl-d7)aminophenazone, Isopyrin-d7, Isopyrine-d7 |
| InChI-Formel | InChI=1S/C14H19N3O/c1-10(2)15-13-11(3)16(4)17(14(13)18)12-8-6-5-7-9-12/h5-10,15H,1-4H3/i1D3,2D3,10D |
| SMILES | [2H]C([2H])([2H])C([2H])(NC1=C(C)N(C)N(C1=O)c2ccccc2)C([2H])([2H])[2H] |
| IUPAC-Name | 4-(1,1,1,2,3,3,3-Heptadeuteriopropan-2-ylamino)-1,5-dimethyl-2-phenylpyrazol-3-on |
| Molekulargewicht (g/mol) | 252.363 |
| Mehr anzeigen |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?