missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAMP3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550254
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total RAMP3 protein.
RAMP3 Polyclonal antibody specifically detects RAMP3 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
RAMP3 | |
Polyclonal | |
Unconjugated | |
Ramp3 | |
AI850306; calcitonin receptor-like receptor activity modifying protein 3; calcitonin-receptor-like receptor activity-modifying protein 3; CRLR activity-modifying protein 3; Ramp3; receptor (calcitonin) activity modifying protein 3; receptor (G protein-coupled) activity modifying protein 3; receptor activity modifying protein 3; receptor activity-modifying protein 3; receptor-activity-modifying protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10268 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O60896 | |
Ramp3 | |
Fusion protein corresponding to a region derived from internal residues of human receptor (G protein-coupled) activity modifying protein 3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |