missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASEF Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 28162-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Binds predomitly GDP, and also GTP.Spezifikation
RASEF | |
Polyclonal | |
Unconjugated | |
RASEF | |
FLJ31614, RAB45, Ras related protein Rab 45, RASEF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
158158, 298138 | |
-20°C | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8IZ41 | |
RASEF | |
RASEF Fusion Protein Ag28049 | |
20 μL | |
Primary | |
Rat, Human | |
Antibody | |
IgG |