missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Klrd1, Rat, FITC, Clone: 18D3, Abnova™
Rat monoclonal antibody raised against Klrd1.
Marke: Abnova MAB5788.500ug
Weitere Details : Netto-Gewicht : 0.00010kg
Spezifikation
Klrd1 | |
Monoclonal | |
FITC | |
Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user. | |
Klrd1 | |
Klrd1 | |
CHO cells transfected with the B6 allele of CD94. | |
RUO | |
16643 | |
Mouse | |
Liquid |
Flow Cytometry | |
18D3 | |
Rat monoclonal antibody raised against Klrd1. | |
In PBS (0.09% sodium azide) | |
CD94 | |
Rat | |
500 μg | |
Primary | |
Mouse CD94 receptor subunit, Mr 70 KDa. | |
Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
IgG2a κ |