Learn More
Invitrogen™ Rat IL-6, Recombinant Protein
Recombinant Protein
Marke:  Invitrogen™ RRIL65
Weitere Details : Netto-Gewicht : 0.00800kg
Artikelnummer. 11868811
  • 267.00 € / 5 Mikrogramm
Verfügbar ab: 15-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.
Beschreibung
Beschreibung
Rat Interleukin 6 (IL-6) Recombinant Protein, RRIL65, from Invitrogen.
Interleukin 6 (IL-6) is a multifunctional 26 kD protein originally discovered in the medium of RNA-stimulated fibroblastoid cells. IL-6 appears to be directly involved in the responses that occur after infection and cellular injury, and it may prove to be important as IL-1 and TNF-a in regulating the acute phase response. IL-6 is reported to be produced by fibroblasts, activated T cells, activated monocytes or macrophages and endothelial cells. IL-6 acts upon a variety of cells including myeloid progenitor cells, T cells, B cells, and hepatocytes. In addition, IL-6 appears to interact with IL-2 in the proliferation of T lymphocytes. IL-6 potentiates the proliferative effect of IL-3 on multipotential hematopoietic progenitors. IL-6 plays a critical role in B-cell differentiation to plasma cells and is a potent growth factor for plasmacytoma and myeloma. IL-6 is a very useful culture supplement for the generation of a high number of antibody-producing hybridomas. Primarily produced at sites of acute and chronic inflammation, IL-6 is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of IL-6 is implicated in a wide variety of inflammation-associated disease states including diabetes mellitus and systemic juvenile rheumatoid arthritis. Purity: >98% pure by SDS-PAGE analyses. Activity: One unit of activity equals 0.01ng/mL, as determined in a proliferation assay with IL-6 dependent mouse 7TD1 cells. Use: Rat IL-6 exerts activity in the range of 0.01 to 1.0ng/mL for most in vitro applications. Reconstitution and Dilution: Reconstitute in distilled water or buffer at pH >7 to a concentration of 100nguL. After reconstitution, dilute cytokine in a neutral buffer containing carrier protein such as 5-10% FBS or 1-4% high purity BSA (fatty acid free fraction V). Lyophilized product is stable until expiration if stored desiccated below 0°C. Cytokines diluted as indicated above should be divided into aliquots and stored at -70°C. Recombinant proteins should be stored at concentrations >500ng/mL to reduce loss of activity.
Spezifikation
Spezifikation
P20607
protein with no preservative; pH >7
Rat IL-6
10μg
-20°C, Avoid Freeze/Thaw Cycles
B-cell hybridoma growth factor; interleukin HP-1; interleukin-6
Il6
Unconjugated
E. coli
Lyophilized
purified
Control, ELISA, Immunoprecipitation, Western Blot
24498
>7
E. Coli
RUO
Rat IL-6
Rat
Rt
Recombinant
Recombinant
Produktvorschläge