missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rb Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 17218-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human RB1.
ZNF639 binds DNA and may function as a transcriptional repressor.Spezifikation
Rb | |
Polyclonal | |
Unconjugated | |
RB1 | |
OSRC, p105 Rb, pp110, pRb, Rb, RB1, retinoblastoma 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5925 | |
-20°C | |
Liquid |
Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P06400 | |
RB1 | |
Rb Fusion Protein Ag11035 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |