missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RBMS3 Polyclonal Antibody

Artikelnummer. 15704931 Alle ansehen Thermo Scientific Produkte
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
15704931 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15704931 Lieferant Invitrogen™ Lieferanten-Nr. PA557028

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

Immunogen sequence: MNHLSLGTTG TIQSQDRIMI LHQLLCQYMT AAAPMQGTYI PQYTPVPPTA VSIEGVVADT SPQTVAPSSQ DTSGQQQQI Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

This gene encodes an RNA-binding protein that belongs to the c-myc gene single-strand binding protein family. These proteins are characterized by the presence of two sets of ribonucleoprotein consensus sequence (RNP-CS) that contain conserved motifs, RNP1 and RNP2, originally described in RNA binding proteins, and required for DNA binding. These proteins have been implicated in such diverse functions as DNA replication, gene transcription, cell cycle progression and apoptosis. The encoded protein was isolated by virtue of its binding to an upstream element of the alpha2(I) collagen promoter. The observation that this protein localizes mostly in the cytoplasm suggests that it may be involved in a cytoplasmic function such as controlling RNA metabolism, rather than transcription. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen RBMS3
Anwendungen Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Konzentration 0.4 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gen RBMS3
Gen-Zugriffsnummer Q6XE24
Gen-Alias 6720477E09Rik; 8430436O14Rik; RBMS3; RNA binding motif single stranded interacting protein 3; RNA binding motif, single stranded interacting protein; RNA binding motif, single stranded interacting protein 3; RNA-binding motif, single-stranded-interacting protein 3; RNA-binding protein; RNA-binding protein RBMS3
Gensymbole RBMS3
Wirtsspezies Rabbit
Immunogen Recombinant protein corresponding to Human RBMS3. Recombinant protein control fragment (Product #RP-96348).
Reinigungsverfahren Antigen affinity chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 27303
Zielspezies Human
Inhalt und Lagerung Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.