missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
RDH10 Polyclonal antibody specifically detects RDH10 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
Specifications
| Antigen | RDH10 |
| Anwendungen | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Klassifikation | Polyclonal |
| Konjugat | Alexa Fluor 350 |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | EC 1.1.1, EC 1.1.1.300, EC 1.1.1.n2, retinol dehydrogenase 10, retinol dehydrogenase 10 (all-trans), SDR16C4, short chain dehydrogenase/reductase family 16C, member 4 |
| Wirtsspezies | Rabbit |
| Immunogen | retinol dehydrogenase 10(all-trans). (Uniprot# Q8IZV5) |
| Reinigungsverfahren | Immunogen Affinity-purified |
| Menge | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?