missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine TOP1 Protein
A cDNA sequence encoding the TOP1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13784-20ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
TOP1 Protein | |
Research Use Only | |
Recombinant Protein | |
Bovine | |
DAKVLKDAKT KKVVESKKKA VQRLEEQLMK LEVQATDREE NKQIALGTSK LNYLDPRITV AWCKKWG | |
Greater than 70% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Bovine | |
Untagged | |
TOP1 is supplied at a concentration of 1 mg/1ml in 16mM HEPES buffer pH 7.5, 400mM NaCl and 20% glycerol. |