Learn More
enQuireBio™ Recombinant Canine MCP 2 Protein
A cDNA sequence encoding the MCP 2 was constructed and used to recombinantly synthesize the protein.
Marke:  enQuireBio™ QP10782-2ug
Weitere Details : Netto-Gewicht : 0.01000kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15925158
Menge Preis
1 129.74 € / 2 Mikrogramm
10 121.33 € / 2 Mikrogramm
Voraussichtliches Versanddatum: 19-01-2022
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Spezifikation
448792
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
CCL8
Recombinant Protein
E. coli
QPDSVSIPIT CCFSMVKRKI PMQKLESYMR ITNSQCPQEA VIFKTKASRE ICADPKQKWV QDYMNHLDQK SQAQKP
MCP 2 Protein
2 μg
< 1.0 EU per ug protein as determined by the LAL method.
The biological activity determined by a chemotaxis bioassay using human peripheral blood monocytes is in a concentration range of 10-100 ng/ml.
Canine
Untagged
MCP-2 protein was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4.