missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Cynomolgus Monkey Fas/TNFRSF6/CD95 Fc, CF

Artikelnummer. 18765966
Change view
Click to view available options
Menge:
50 μg
Packungsgröße:
50 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18765966 50 μg 50 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18765966 Lieferant R&D Systems™ Lieferanten-Nr. 10755FS050

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

Measured by its ability to inhibit Fas Ligand-induced apoptosis of Jurkat human acute T cell leukemia cells. Cheng, J. et al. (1994) Science 263:1759. The ED50 for this effect is 40.0-240 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Zugriffsnummer Q9TSN4.1
Zur Verwendung mit (Anwendung) Bioactivity
Zusammensetzung Lyophilized from a 0.2 μm filtered solution in PBS.
Gen-ID (Entrez) 355
Molekulargewicht MolecularWeight-observed: 55-70 kDa, under reducing conditions, MolecularWeight-theroretical: 43 kDa
Menge 50 μg
Quelle Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey Fas/TNFRSF6/CD95 protein Cynomolgus Monkey Fas/TNFRSF6/CD95 (Gln26-Asp173) (N-terminus) Accession # Q9TSN4.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Lagerungsbedingungen Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin-Konzentration <0.10 EU per 1 μg of the protein by the LAL method.
Gen-Alias Apo-1, Apo-1 antigen, apoptosis antigen 1, Apoptosis-mediating surface antigen FAS, APT1, APT1FASTM, CD95, CD95 antigen, CD95ALPS1A, Fas (TNF receptor superfamily, member 6), Fas AMA, Fas antigen, FAS1, FASLG receptor, TNFRSF6, TNFRSF6member 6, tumor necrosis factor receptor superfamily member 6
Konjugat Unconjugated
Rekonstitution Reconstitute at 500 μg/mL in PBS.
Reinheits- oder Qualitätsgrad >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Fas/TNFRSF6/CD95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.