missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Cytomegalovirus Uncharacterized protein UL128 Protein
A DNA sequence encoding the Human cytomegalovirus Uncharacterized protein UL128, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP9537-ye-500ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
Uncharacterized protein UL128 Protein | |
Research Use Only | |
UL128 | |
Recombinant Protein | |
Yeast | |
MSPKDLTPFL TTLWLLLGHS RVPRVRAEEC CEFINVNHPP ERCYDFKMCN RFTVALRCPD GEVCYSPEKT AEIRGIVTTM THSLTRQVVH NKLTSCNYNP LYLEADGRIR CGKVNDKAQY LLGAAGSVPY RWINLEYDKI TRIVGLDQYL ESVKKHKRLD VCRAKMGYML Q | |
Greater than 90% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Full Length | |
Cytomegalovirus | |
His | |
Tris-based buffer, 50% glycerol |