Learn More
enQuireBio™ Recombinant Dog IL-8 / CXCL8 Protein
A DNA sequence encoding the Canis familiaris (Dog) (Canis lupus familiaris) IL-8 / CXCL8, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP6227-ec-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_001003200.1 | |
IL-8 / CXCL8 Protein | |
1 mg | |
Not determined. | |
Full Length | |
Dog | |
GST | |
Tris-based buffer, 50% glycerol |
403850 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
CXCL8 | |
Recombinant Protein | |
E. coli | |
AVLSRVSSEL RCQCIKTHST PFHPKYIKEL RVIDSGPHCE NSEIIVKLFN GNEVCLDPKE KWVQKVVQIF LKKAEKQDP |