missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. Coli GLDA Protein
A cDNA sequence encoding the GLDA was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10644-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
948440 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity: > 14 Units/ml. One unit will oxidize 1.0 umole of glycerol to dihydroxyacetone per minute at pH 8.0 at 25C. | |
E. coli | |
His | |
GLDA protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4) and 10% glycerol. |
GLDA Protein | |
Research Use Only | |
gldA | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDRIIQS PGKYIQGADV INRLGEYLKP LAERWLVVGD KFVLGFAQST VEKSFKDAGL VVEIAPFGGE CSQNEIDRLR GIAETAQCGA ILGIGGGKTL DTAKALAHFM GVPVAIAPTI ASTDAPCSAL SVIYTDEGEF DRYLLLPNNP NMVIVDTKIV AGAPARLLAA GIGDALATWF EARACSRSGA TTMAGGKCTQ AALALAELCY NTLLEEGEKA MLAAEQHVVT PALERVIEAN TYLSGVGFES GGLAAAHAVH NGLTAIPDAH HYYHGEKVAF GTLTQLVLEN APVEEIETVA ALSHAVGLPI TLAQLDIKED VPAKMRIVAE AACAEGETIH NMPGGATPDQ VYAALLVADQ YGQRFLQEWE | |
Greater than 95.0% as determined by SDS-PAGE. |