missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS4 Protein
A cDNA sequence encoding the HCV NS4 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12176-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
Hepatitis C HCV NS4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-HCl pH 8.0, 1mM EDTA, 1.5M urea and 50%glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
Untagged | |
HCV NS4 Protein is >95% pure as determined by 10% PAGE (coomassie staining). |