missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HSV HCV Core 1-120 Protein
A cDNA sequence encoding the HCV Core 1-120 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12139-5ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
HSV HCV Core 1-120 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSTNPKPQRK TKRNTNRRPQ DVKFPGGGQI VGGVYLLPRR GPRLGVRATR KTSERSQPRG RRQPIPKARR PEGRTWAQPG YPWPLYGNEG CGWAGWLLSP RGSRPSWGPT DPRRRSRNLG | |
Greater than 85% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HSV | |
His | |
The HCV Core protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |