missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACO1 Protein
A cDNA sequence encoding the ACO1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10940-50ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
ACO1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSNPFAH LAEPLDPVQP GKKFFNLNKL EDSRYGRLPF SIRVLLEAAI RNCDEFLVKK QDIENILHWN VTQHKNIEVP FKPARVILQD FTGVPAVVDF AAMRDAVKKL GGDPEKINPV CPADLVIDHS IQVDFNRRAD SLQKNQDLEF ERNRERFEFL KWGSQAFHNM RIIPPGSGII HQVNLEYLAR VVFDQDGYYY PDSLVGTDSH TTMIDGLGIL GWGVGGIEAE AVMLGQPISM VLPQVIGYRL MGKPHPLVTS TDIVLTITKH LRQVGVVGKF VEFFGPGVAQ LSIADRATIA NMCPEYGATA AFFPVDEVSI TYLVQTGRDE EKLKYIKKYL QAVGMFRDFN DPSQDPDFTQ VVELDLKTVV PCCSGPKRPQ DKVAVSDMKK DFESCLGAKQ GFKGFQVAPE HHNDHKTFIY DNTEFTLAHG SVVIAAITSC TNTSNPSVML GAGLLAKKAV DAGLNVMPYI KTSLSPGSGV VTYYLQESGV MPYLSQLGFD VVGYGCMTCI GNSGPLPEPV VEAITQGDLV AVGVLSGNRN FEGRVHPNTR ANYLASPPLV IAYAIAGTIR IDFEKEPLGV NAKGQQVFLK DIWPTRDEIQ AVERQYVIPG MFKEVYQKIE TVNESWNALA TPSDKLFFWN SKSTYIKSPP FFENLTLDLQ PPKSIVDAYV LLNLGDSVTT DHISPAGNIA RNSPAARYLT NRGLTPREFN SYGSRRGNDA VMARGTFANI RLLNRFLNKQ APQTIHLPSG EILDVFDAAE RYQQAGLPLI VLAGKEYGAG SSRDWAAKGP FLLGIKAVLA ESYERIHRSN LVGMGVIPLE YLPGENADAL GLTGQERYTI IIPENLKPQM KVQVKLDTGK TFQAVMRFDT DVELTYFLNG GILNYMIRKM | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ACO1 protein solution (0.5 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0)2mM DTT, 100mM NaCl and 10% glycerol. |