missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ALAD Protein
A cDNA sequence encoding the ALAD was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10996-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
210 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMQPQSV LHSGYFHPLL RAWQTATTTL NASNLIYPIF VTDVPDDIQP ITSLPGVARY GVKRLEEMLR PLVEEGLRCV LIFGVPSRVP KDERGSAADS EESPAIEAIH LLRKTFPNLL VACDVCLCPY TSHGHCGLLS ENGAFRAEES RQRLAEVALA YAKAGCQVVA PSDMMDGRVE AIKEALMAHG LGNRVSVMSY SAKFASCFYG PFRDAAKSSP AFGDRRCYQL PPGARGLALR AVDRDVREGA DMLMVKPGMP YLDIVREVKD KHPDLPLAVY HVSGEFAMLW HGAQAGAFDL KAAVLEAMTA FRRAGADIII TYYTPQLLQW LKEE | |
Greater than 85% as determined by SDS-PAGE. |
ALAD Protein | |
Research Use Only | |
ALAD | |
Human | |
His | |
The ALAD solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl and 10% glycerol. |