missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Arylamine N-acetyltransferase 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Arylamine N-acetyltransferase 2, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP6405-ec-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_000006.2 | |
Arylamine N-acetyltransferase 2 Protein | |
Research Use Only | |
NAT2 | |
Recombinant Protein | |
E. coli | |
MDIEAYFERI GYKNSRNKLD LETLTDILEH QIRAVPFENL NMHCGQAMEL GLEAIFDHIV RRNRGGWCLQ VNQLLYWALT TIGFQTTMLG GYFYIPPVNK YSTGMVHLLL QVTIDGRNYI VDAGSGSSSQ MWQPLELISG KDQPQVPCIF CLTEERGIWY LDQIRREQYI TNKEFLNSHL LPKKKHQKIY LFTLEPRTIE DFESMNTYLQ TSPTSSFITT SFCSLQTPEG VYCLVGFILT YRKFNYKDNT DLVEFKTLTE EEVEEVLKNI FKISLGRNLV PKPGDGSLTI | |
Greater than 90% as determined by SDS-PAGE. |
10 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |