missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AURKB Protein
A cDNA sequence encoding the AURKB was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11113-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
9212 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAQKENSYPW PYGRQTAPSG LSTLPQRVLR KEPVTPSALV LMSRSNVQPT AAPGQKVMEN SSGTPDILTR HFTIDDFEIG RPLGKGKFGN VYLAREKKSH FIVALKVLFK SQIEKEGVEH QLRREIEIQA HLHHPNILRL YNYFYDRRRI YLILEYAPRG ELYKELQKSC TFDEQRTATI MEELADALMY CHGKKVIHRD IKPENLLLGL KGELKIADFG WSVHAPSLRR KTMCGTLDYL PPEMIEGRMH NEKVDLWCIG VLCYELLVGN PPFESASHNE TYRRIVKVDL KFPASVPMGA QDLISKLLRH NPSERLPLAQ VSAHPWVRAN SRRVLPPSAL QSVA | |
Greater than 90.0% as determined by SDS-PAGE. |
AURKB Protein | |
Research Use Only | |
AURKB | |
Human | |
His | |
The AURKB solution containing 20mM Tris-HCl buffer (pH 8.0), 0.5mM DTT, 20% glycerol, 0.1mM EDTA, 0.1mM EGTA, 0.1M NaCl and 0.1mM PMSF. |