missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BPNT1 Protein
A cDNA sequence encoding the BPNT1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11199-10ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
BPNT1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMASS NTVLMRLVAS AYSIAQKAGM IVRRVIAEGD LGIVEKTCAT DLQTKADRLA QMSICSSLAR KFPKLTIIGE EDLPSEEVDQ ELIEDSQWEE ILKQPCPSQY SAIKEEDLVV WVDPLDGTKE YTEGLLDNVT VLIGIAYEGK AIAGVINQPY YNYEAGPDAV LGRTIWGVLG LGAFGFQLKE VPAGKHIITT TRSHSNKLVT DCVAAMNPDA VLRVGGAGNK IIQLIEGKAS AYVFASPGCK KWDTCAPEVI LHAVGGKLTD IHGNVLQYHK DVKHMNSAGV LATLRNYDYY ASRVPESIKN ALVP | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The BPNT1 protein solution (0.5 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0) 5mM DTT, 0.1M NaCl and 10% glycerol. |