missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BTN3A1 / CD277 Protein
A DNA sequence encoding the Homo sapiens (Human) BTN3A1 / CD277, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP5729-ye-200ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_001138480.1 | |
BTN3A1 / CD277 Protein | |
Research Use Only | |
BTN3A1 | |
Recombinant Protein | |
Yeast | |
QFSVLGPSGP ILAMVGEDAD LPCHLFPTMS AETMELKWVS SSLRQVVNVY ADGKEVEDRQ SAPYRGRTSI LRDGITAGKA ALRIHNVTAS DSGKYLCYFQ DGDFYEKALV ELKVAALGSD LHVDVKGYKD GGIHLECRST GWYPQPQIQW SNNKGENIPT VEAPVVADGV GLYAVAASVI MRGSSGEGVS CTIRSSLLGL EKTASISIAD PFFRSAQRWI AALAG | |
Greater than 90% as determined by SDS-PAGE. |
11119 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |