missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CAB39L Protein
A cDNA sequence encoding the CAB39L was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11250-5ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
CAB39L Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMKKMPLF SKSHKNPAEI VKILKDNLAI LEKQDKKTDK ASEEVSKSLQ AMKEILCGTN EKEPPTEAVA QLAQELYSSG LLVTLIADLQ LIDFEGKKDV TQIFNNILRR QIGTRSPTVE YISAHPHILF MLLKGYEAPQ IALRCGIMLR ECIRHEPLAK IILFSNQFRD FFKYVELSTF DIASDAFATF KDLLTRHKVL VADFLEQNYD TIFEDYEKLL QSENYVTKRQ SLKLLGELIL DRHNFAIMTK YISKPENLKL MMNLLRDKSP NIQFEAFHVF KVFVASPHKT QPIVEILLKN QPKLIEFLSS FQKERTDDEQ FADEKNYLIK QIRDLKKTAP | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CAB39L protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl,10% glycerol and 1mM DTT. |