missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDK2AP2 Protein
A cDNA sequence encoding the CDK2AP2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11374-20ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
10263 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSYKPIA PAPSSTPGSS TPGPGTPVPT GSVPSPSGSV PGAGAPFRPL FNDFGPPSMG YVQAMKPPGA QGSQSTYTDL LSVIEEMGKE IRPTYAGSKS AMERLKRGII HARALVRECL AETERNART | |
Greater than 95.0% as determined by SDS-PAGE. |
CDK2AP2 Protein | |
Research Use Only | |
CDK2AP2 | |
Human | |
His | |
CDK2AP2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol and 2mM DTT. |