missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CYTL1 Protein
A cDNA sequence encoding the CYTL1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11593-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
54360 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASTPPTCYSRMR ALSQEITRDF NLLQVSEPSE PCVRYLPRLY LDIHNYCVLD KLRDFVASPP CWKVAQVDSL KDKARKLYTI MNSFCRRDLV FLLDDCNALE YPIPVTTVLP DRQR | |
Greater than 95.0% as determined by SDS-PAGE. |
CYTL1 Protein | |
Research Use Only | |
CYTL1 | |
Human | |
His | |
CYTL1 was filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 20mM Tris buffer and 50mM NaCl, pH 7.5. |