missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DCUN1D1 / SCCRO Protein
A DNA sequence encoding the Homo sapiens (Human) DCUN1D1 / SCCRO, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP7895-ec-100ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
NP_001295030.1 | |
DCUN1D1 / SCCRO Protein | |
Research Use Only | |
DCUN1D1 | |
Recombinant Protein | |
E. coli | |
MNKLKSSQKD KVRQFMIFTQ SSEKTAVSCL SQNDWKLDVA TDNFFQNPEL YIRESVKGSL DRKKLEQLYN RYKDPQDENK IGIDGIQQFC DDLALDPASI SVLIIAWKFR AATQCEFSKQ EFMDGMTELG CDSIEKLKAQ IPKMEQELKE PGRFKDFYQF TFNFAKNPGQ KGLDLEMAIA YWNLVLNGRF KFLDLWNKFL LEHHKRSIPK DTWNLLLDFS TMIADDMSNY DEEGAWPVLI DDFVEFARPQ IAGTKSTTV | |
Greater than 90% as determined by SDS-PAGE. |
54165 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |