missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human F-box / LRR-repeat protein 1
A DNA sequence encoding the Homo sapiens (Human) F-box/LRR-repeat protein 2, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP8251-ec-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_001165184.1 | |
F-box / LRR-repeat protein 1 | |
Research Use Only | |
FBXL2 | |
Recombinant Protein | |
E. coli | |
MVFSNNDEGL INKKLPKELL LRIFSFLDIV TLCRCAQISK AWNILALDGS NWQRIDLFNF QTDVEGRVVE NISKRCGGFL RKLSLRGCIG VGDSSLKTFA QNCRNIEHLN LNGCTKITDS TCYSLSRFCS KLKHLDLTSC VSITNSSLKG ISEGCRNLEY LNLSWCDQIT KDGIEALVRG CRGLKALLLR GCTQLEDEAL KHIQNYCHEL VSLNLQSCSR ITDEGVVQIC RGCHRLQALC LSGCSNLTDA SLTALGLNCP RLQILEAARC SHLTDAGFTL LARNCHELEK MDLEECILIT DSTLIQLSIH CPKLQALSLS HCELITDDGI LHLSNSTCGH ERLRVLELDN CLLITDVALE HLENCRGLER LELYDCQQVT RAGIKRMRAQ LPHVKVHAYF APVTPPTAVA GSGQRLCRCC VIL | |
Greater than 90% as determined by SDS-PAGE. |
25827 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |