Learn More
enQuireBio™ Recombinant Human Gem-associated protein 1
A DNA sequence encoding the Homo sapiens (Human) Gem-associated protein 7, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP8052-ec-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_001007270.1 | |
Gem-associated protein 1 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
79760 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
GEMIN7 | |
Recombinant Protein | |
E. coli | |
MQTPVNIPVP VLRLPRGPDG FSRGFAPDGR RAPLRPEVPE IQECPIAQES LESQEQRARA ALRERYLRSL LAMVGHQVSF TLHEGVRVAA HFGATDLDVA NFYVSQLQTP IGVQAEALLR CSDIISYTFK P |