missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HbA1c Protein
A cDNA sequence encoding the HbA1c was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12117-10mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
HbA1c Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
The HbA1c is supplied in a proprietary buffer formulation having pH 8.0. The HbA1c was dispensed on the basis of HbA1c not total hemoglobin. |
10 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Greater than 96.0%. |