missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSBP1L1 Protein
A cDNA sequence encoding the HSBP1L1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12318-5ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
HSBP1L1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDVRGPE APGGRALRDA AENLFQELQE HFQALTATLN LRMEEMGNRI EDLQKNVNDL MVQAGIENSI KEQMLKT | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
HSBP1L1 protein solution (1 mg/ml) containing Phosphate buffer saline (pH 7.4) and 10% glycerol. |