missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IFI30 Protein
A cDNA sequence encoding the IFI30 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12372-10ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
10437 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMNAPLVN VTLYYEALCG GCRAFLIREL FPTWLLVMEI LNVTLVPYGN AQEQNVSGRW EFKCQHGEEE CKFNKVEACV LDELDMELAF LTIVCMEEFE DMERSLPLCL QLYAPGLSPD TIMECAMGDR GMQLMHANAQ RTDALQPPHE YVPWVTVNGK PLEDQTQLLT LVCQLYQGK | |
Greater than 90% as determined by SDS-PAGE. |
IFI30 Protein | |
Research Use Only | |
IFI30 | |
Human | |
His | |
The IFI30 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 10% glycerol. |