missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IGF1 Protein
A cDNA sequence encoding the IGF1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10680-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
3479 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by a cell proliferation assay using murine BALB/C 3T3 cells is less than 1.0 ng/ml, corresponding to a specific activity of > 1M 106 units/mg. | |
Human | |
Untagged | |
The protein was lyophilized from a 0.2microm filtered concentrated solution in 20mM PBS, pH 7.0, 130mM NaCl. |
IGF1 Protein | |
Research Use Only | |
IGF1 | |
Recombinant Protein | |
E. coli | |
GPETLCGAEL VDALQFVCGD RGFYFNKPTG YGSSSRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPAKSA | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC(b) Analysis by SDS-PAGE. |