missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IGFBP 4 Protein
A cDNA sequence encoding the IGFBP 4 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10684-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
IGFBP 4 Protein | |
Research Use Only | |
The ED50 range is 0.01-0.09microg/ml as measured by its ability to inhibit the biological activity of IGFII (20ng/ml) on MCF7 human breast adenocarcinoma cells. | |
Human | |
His | |
IGFBP 4 was lyophilized after extensive dialysis against PBS. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Human (HEK293) | |
DEAIHCPPCS EEKLARCRPP VGCEELVREP GCGCCATCAL GLGMPCGVYT PRCGSGLRCY PPRGVEKPLH TLMHGQGVCM ELAEIEAIQE SLQPSDKDEG DHPNNSFSPC SAHDRRCLQK HFAKIRDRST SGGKMKVNGA PREDARPVPQ GSCQSELHRA LERLAASQSR THEDLYIIPI PNCDRNGNFH PKQCHPALDG QRGKCWCVDR KTGVKLPGGL EPKGELDCHQ LADSFRE + His Tag | |
Greater than 95.0% as determined by SDS-PAGE. |