missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL12RB1 Protein
A cDNA sequence encoding the IL12RB1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12404-HIS-INSECT-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
3594 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
MGSSHHHHHH SSGLVPRGSH MGSMEPLVTW VVPLLFLFLL SRQGAACRTS ECCFQDPPYP DADSGSASGP RDLRCYRISS DRYECSWQYE GPTAGVSHFL RCCLSSGRCC YFAAGSATRL QFSDQAGVSV LYTVTLWVES WARNQTEKSP EVTLQLYNSV KYEPPLGDIK VSKLAGQLRM EWETPDNQVG AEVQFRHRTP SSPWKLGDCG PQDDDTESCL CPLEMNVAQE FQLRRRQLGS QGSSWSKWSS PVCVPPENPP QPQVRFSVEQ LGQDGRRRLT LKEQPTQLEL PEGCQGLAPG TEVTYRLQLH MLSCPCKAKA TRTLHLGKMP YLSGAAYNVA VISSNQFGPG LNQTWHIPAD THTEPVALNI SVGTNGTTMY WPARAQSMTY CIEWQPVGQD GGLATCSLTA PQDPDPAGMA TYSWSRESGA MGQEKCYYIT IFASAHPEKL TLWSTVLSTY HFGGNASAAG TPHHVSVKNH SLDSVSVDWA PSLLSTCPGV LKEYVVRCRD EDSKQVSEHP VQPTETQVTL SGLRAGVAYT VQVRADTAWL RGVWSQPQRF SIEVQVSD | |
Greater than 90.0% as determined by SDS-PAGE. |
IL12RB1 Protein | |
Research Use Only | |
IL12RB1 | |
Human | |
His | |
IL12RB1 protein solution (0.25 mg/ml) containing 20mM TRIS 8.0 buffer and 10% glycerol. |