missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LSM1 Protein
A cDNA sequence encoding the LSM1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12589-5ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
LSM1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MNYMPGTASL IEDIDKKHLV LLRDGRTLIG FLRSIDQFAN LVLHQTVERI HVGKKYGDIP RGIFVVRGEN VVLLGEIDLE KESDTPLQQV SIEEILEEQR VEQQTKLEAE KLKVQALKDR GLSIPRADTL DEY | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The LSM1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 7.5), 1mM DTT, 10% glycerol and 0.1M NaCl. |