missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LSM5 Protein
A cDNA sequence encoding the LSM5 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12593-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
LSM5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAANATT NPSQLLPLEL VDKCIGSRIH IVMKSDKEIV GTLLGFDDFV NMVLEDVTEF EITPEGRRIT KLDQILLNGN NITMLVPGGE GPEV | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
LSM5 protein solution (1 mg/ml) containing 20mM Tris-HCl (pH 8.5), 10% glycerol, 0.15M NaCl and 1mM DTT. |