missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MET Protein
A cDNA sequence encoding the MET was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12683-2ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
4233 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
MKAPAVLAPG ILVLLFTLVQ RSNGECKEAL AKSEMNVNMK YQLPNFTAET PIQNVILHEH HIFLGATNYI YVLNEEDLQK VAEYKTGPVL EHPDCFPCQD CSSKANLSGG VWKDNINMAL VVDTYYDDQL ISCGSVNRGT CQRHVFPHNH TADIQSEVHC IFSPQIEEPS QCPDCVVSAL GAKVLSSVKD RFINFFVGNT INSSYFPDHP LHSISVRRLK ETKDGFMFLT DQSYIDVLPE FRDSYPIKYV HAFESNNFIY FLTVQRETLD AQTFHTRIIR FCSINSGLHS YMEMPLECIL TEKRKKRSTK KEVFNILQAA YVSKPGAQLA RQIGASLNDD ILFGVFAQSK PDSAEPMDRS AMCAFPIKYV NDFFNKIVNK NNVRCLQHFY GPNHEHCFNR TLLRNSSGCE ARRDEYRTEF TTALQRVDLF MGQFSEVLLT SISTFIKGDL TIANLGTSEG RFMQVVVSRS GPSTPHVNFL LDSHPVSPEV IVEHTLNQNG YTLVITGKKI TKIPLNGLGC RHFQSCSQCL SAPPFVQCGW CHDKCVRSEE CLSGTWTQQI CLPAIYKVFP NSAPLEGGTR LTICGWDFGF RRNNKFDLKK TRVLLGNESC TLTLSESTMN TLKCTVGPAM NKHFNMSIII SNGHGTTQYS TFSYVDPVIT SISPKYGPMA GGTLLTLTGN YLNSGNSRHI SIGGKTCTLK SVSNSILECY TPAQTISTEF AVKLKIDLAN RETSIFSYRE DPIVYEIHPT KSFISGGSTI TGVGKNLNSV SVPRMVINVH EAGRNFTVAC QHRSNSEIIC CTTPSLQQLN LQLPLKTKAF FMLDGILSKY FDLIYVHNPV FKPFEKPVMI SMGNENVLEI KGNDIDPEAV KGEVLKVGNK SCENIHLHSE AVLCTVPNDL LKLNSELNIE |
MET Protein | |
Research Use Only | |
MET | |
Human | |
Untagged | |
MET is supplied in 100microg/ml in 50mM Tris-HCl (pH 7.5) with 10mM L-glutathione (reduced). |